Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trifluoro-6-nitroaniline, 98%, Thermo Scientific™

Catalog No. AAH6151403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6151403 1 g
AAH6151406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6151403 Supplier Thermo Scientific Chemicals Supplier No. H6151403

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148416-38-0
Molecular Formula C6H3F3N2O2
Molecular Weight (g/mol) 192.097
MDL Number MFCD00174089
InChI Key ILENVKAVEFKZSD-UHFFFAOYSA-N
Synonym 6-nitro-2,3,4-trifluoroaniline, 2-nitro-4,5,6-trifluoroaniline, pubchem4504, acmc-1bzfn, intermediates-zcf02138, ksc525q0b, attercop-chm at116509, buttpark 24\01-62, 2,3,4-trifluoro-6-nitro-aniline, 2,3,4-trifluoro-6-nitrophenylamine
PubChem CID 2775763
IUPAC Name 2,3,4-trifluoro-6-nitroaniline
SMILES C1=C(C(=C(C(=C1F)F)F)N)[N+](=O)[O-]

Specifications

Melting Point 59°C to 61°C
Color White
Quantity 1 g
Formula Weight 192.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,4-Trifluoro-6-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.