Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trifluoro-6-nitroaniline 98.0+%, TCI America™
SDP

Catalog No. T27835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27835G 5 g
T27831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27835G Supplier TCI America Supplier No. T27835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 148416-38-0
Molecular Formula C6H3F3N2O2
Molecular Weight (g/mol) 192.097
MDL Number MFCD00174089
InChI Key ILENVKAVEFKZSD-UHFFFAOYSA-N
Synonym 6-nitro-2,3,4-trifluoroaniline, 2-nitro-4,5,6-trifluoroaniline, pubchem4504, acmc-1bzfn, intermediates-zcf02138, ksc525q0b, attercop-chm at116509, buttpark 24\01-62, 2,3,4-trifluoro-6-nitro-aniline, 2,3,4-trifluoro-6-nitrophenylamine
PubChem CID 2775763
IUPAC Name 2,3,4-trifluoro-6-nitroaniline
SMILES C1=C(C(=C(C(=C1F)F)F)N)[N+](=O)[O-]

Specifications

Melting Point 60°C
Quantity 5 g
Formula Weight 192.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trifluoro-6-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.