Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trifluorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. T23051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23051G Supplier TCI America Supplier No. T23051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143879-80-5
Molecular Formula C7H2F3N
Molecular Weight (g/mol) 157.10
MDL Number MFCD00013288
InChI Key KTPHYLJFAZNALV-UHFFFAOYSA-N
Synonym 2,3,4-difluorobenzonitrile, benzonitrile, trifluoro, 2,3,4-trifluorobenzenecarbonitrile, trifluorobenzonitrile, pubchem1572, acmc-209cri, ksc494g9l, 2,3,4-trifluoro benzonitrile, 2,3,4-trifluoro-benzonitrile, benzonitrile, 2,3,4-trifluoro
PubChem CID 518936
IUPAC Name 2,3,4-trifluorobenzonitrile
SMILES FC1=CC=C(C#N)C(F)=C1F

Specifications

Color Yellow
Boiling Point 118°C
Quantity 1 g
UN Number 3276
Formula Weight 157.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4-Trifluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.