Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trifluorotoluene 97.0+%, TCI America™
SDP

Catalog No. T27355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27355G 5 g
T27351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27355G Supplier TCI America Supplier No. T27355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 193533-92-5
Molecular Formula C7H5F3
Molecular Weight (g/mol) 146.112
MDL Number MFCD00075194
InChI Key LRQPEHJWTXCLQY-UHFFFAOYSA-N
PubChem CID 2777991
IUPAC Name 1,2,3-trifluoro-4-methylbenzene
SMILES CC1=C(C(=C(C=C1)F)F)F

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 g
UN Number 1993
Synonym 2,3,4-trifluorotoluene, benzene, 1,2,3-trifluoro-4-methyl, triflurotoluene, 1,2,3-trifluoro-4-methyl-benzene, pubchem1628, acmc-1cn3t, ksc541e8d, benzene,1,2,3-trifluoro-4-methyl, benzene, 1,2,3-trifluoro-4-methyl-9ci
Formula Weight 146.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4-Trifluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.