Learn More
Medchemexpress LLC 2,3,4-trihydroxybenzoic acid | 610-02-6 | MFCD00002447 | 99.8% | 170.12 g/mol | C7H6O5 | 1 ML
SDP
Catalog No. 5000299881
Encompass_Preferred
2,3,4-Trihydroxybenzoic acid is a high-purity analytical standard used as an internal standard for separation of phenolic acids by HPLC. Supplied as a 10 mM solution in 1 mL DMSO for research use, it provides consistent, ready-to-use concentration for method development and quantitation.
  • High purity (99.79%) for accurate quantitation.
  • Supplied as a 10 mM solution in 1 mL DMSO.
  • Suitable as an internal standard for HPLC separation of phenolic acids.
  • Recommended storage: solution at -80°C (up to 6 months) or -20°C (up to 1 month) under nitrogen.
  • Useful for analytical method development and quality control.
Catalog No. 50-002-99881 Supplier Medchemexpress LLC Supplier No. HYW01699310MM/1ML
Add to Cart
Add to Cart
2,3,4-Trihydroxybenzoic acid is a high-purity analytical standard used as an internal standard for separation of phenolic acids by HPLC. Supplied as a 10 mM solution in 1 mL DMSO for research use, it provides consistent, ready-to-use concentration for method development and quantitation.
  • High purity (99.79%) for accurate quantitation.
  • Supplied as a 10 mM solution in 1 mL DMSO.
  • Suitable as an internal standard for HPLC separation of phenolic acids.
  • Recommended storage: solution at -80°C (up to 6 months) or -20°C (up to 1 month) under nitrogen.
  • Useful for analytical method development and quality control.