Learn More
Cayman Chemical 2,3,5,4'-Tetrahydroxyl-diphenylethylene 2-O-β-D-Glucoside, Purity: ≥98%, CAS: 82373-94-2, 100 mg
SDP
Catalog No. 502948141
Encompass_Preferred
A stilbene glucoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 40 µM); inhibits doxorubicin-induced apoptosis, loss of mitochondrial membrane potential, and generation of ROS in mouse cardiomyocytes; increases acetylcholine-induced relaxation of, and NO levels in, aortic rings isolated from a rat model of diet-induced atherosclerosis at 30, 60, and 120 mg/kg; reduces infarct area and neuronal apoptosis in a mouse model of MCAO-induced ischemia and reperfusion injury at 15a
Catalog No. 50-294-8141 Supplier Cayman Chemical Supplier No. 34594100 MG
A stilbene glucoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 40 µM); inhibits doxorubicin-induced apoptosis, loss of mitochondrial membrane potential, and generation of ROS in mouse cardiomyocytes; increases acetylcholine-induced relaxation of, and NO levels in, aortic rings isolated from a rat model of diet-induced atherosclerosis at 30, 60, and 120 mg/kg; reduces infarct area and neuronal apoptosis in a mouse model of MCAO-induced ischemia and reperfusion injury at 15a