Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrabromothieno[3,2-b]thiophene 98.0+%, TCI America™
SDP

Catalog No. T27721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27721G Supplier TCI America Supplier No. T27721G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124638-53-5
Molecular Formula C6Br4S2
Molecular Weight (g/mol) 455.80
MDL Number MFCD03788229
InChI Key CWCKRNKYWPNOAZ-UHFFFAOYSA-N
PubChem CID 2762568
IUPAC Name tetrabromothieno[3,2-b]thiophene
SMILES BrC1=C(Br)C2=C(S1)C(Br)=C(Br)S2

Specifications

Melting Point 234°C
Color Yellow
Quantity 1 g
Formula Weight 455.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrabromothieno[3,2-b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.