Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrachloronitrobenzene 98.0+%, TCI America™
SDP

Catalog No. T0624500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0624500G 500 g
T062425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0624500G Supplier TCI America Supplier No. T0624500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117-18-0
Molecular Formula C6HCl4NO2
Molecular Weight (g/mol) 260.879
MDL Number MFCD00007066
InChI Key XQTLDIFVVHJORV-UHFFFAOYSA-N
Synonym tecnazene, tecnazen, 2,3,5,6-tetrachloronitrobenzene, arena, folosan, fusarex, myfusan, hickstor, turbostore, easytec
PubChem CID 8330
ChEBI CHEBI:82044
IUPAC Name 1,2,4,5-tetrachloro-3-nitrobenzene
SMILES C1=C(C(=C(C(=C1Cl)Cl)[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 100°C
Quantity 500 g
UN Number 3077
Formula Weight 260.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrachloronitrobenzene
RTECSNumber DC0175000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.