Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane

Catalog No. 37677925MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-677-925MG 25 mg
37-677-9100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-677-925MG Supplier Sigma Aldrich Supplier No. 37677925MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane

Specifications

CAS 29261-33-4
Melting Point 285°C to 290°C
Molecular Formula C12F4N4
Linear Formula C12F4N4
MDL Number MFCD00042382
Quantity 25 mg
Synonym (2,3,5,6-Tetrafluoro-2,5-cyclohexadiene-1,4-diylidene)dimalononitrile; 7,7,8,8-Tetracyano-2,3,5,6-tetrafluoroquinodimethane; F4TCNQ
Molecular Weight (g/mol) 276.15
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.