Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluoroaniline 97.0+%, TCI America™
SDP

Catalog No. T10285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10285G 5 g
T102825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10285G Supplier TCI America Supplier No. T10285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 700-17-4
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD00007644
InChI Key SPSWJTZNOXMMMV-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoraniline, 2,3,5,6-tetrafluorobenzenamine, benzenamine, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorophenylamine, acmc-1bm33, 2,3,5, 6-tetrafluoroaniline, 2,3,5,6-tetrafluoro-aniline, 2,3,5,6-tetrafluoro aniline, timtec-bb sbb028353, aniline, 2,3,5,6-tetrafluoro
PubChem CID 12786
IUPAC Name 2,3,5,6-tetrafluoroaniline
SMILES C1=C(C(=C(C(=C1F)F)N)F)F

Specifications

Color White-Yellow
Boiling Point 158°C
Quantity 5 g
Formula Weight 165.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,5,6-Tetrafluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.