Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluorobenzeneboronic acid, 99%

Catalog No. AAL1982603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1982603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1982603 Supplier Thermo Scientific Chemicals Supplier No. L1982603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 511295-01-5 | C6H3BF4O2 | 193.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 511295-01-5
Molecular Formula C6H3BF4O2
Molecular Weight (g/mol) 193.89
MDL Number MFCD04039310
InChI Key SORPNEUIIZJCBF-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorophenyl boronic acid, 2,3,5,6-tetrafluorobenzeneboronic acid, boronic acid, 2,3,5,6-tetrafluorophenyl, boronic acid, b-2,3,5,6-tetrafluorophenyl, acmc-209ks0, 2,3,5,6-tetrafluorophenylboranic acid, 2,3,5,6-tetrakis fluoranyl phenyl boronic acid, boronic acid, 2,3,5,6-tetrafluorophenyl-9ci
PubChem CID 2783275
IUPAC Name (2,3,5,6-tetrafluorophenyl)boronic acid
SMILES OB(O)C1=C(F)C(F)=CC(F)=C1F

Specifications

Melting Point ∼140°C to 145°C
Quantity 1 g
Beilstein 9200167
Formula Weight 193.9
Percent Purity 99%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.