Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluorophenol, 97%

Catalog No. AAA1268606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1268606 5 g
AAA1268614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1268606 Supplier Thermo Scientific Chemicals Supplier No. A1268606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 769-39-1 | C6H2F4O | 166.075 g/mol

2,3,5,6-Tetrafluorophenol is used as a cytochrome P450 substrate. 2,3,5,6-Tetrafluorophenol was used in preparation of radioiodinated phenylalanine derivative which is useful in peptide synthesis, technetium-99m labeled antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5,6-Tetrafluorophenol is used as a cytochrome P450 substrate. 2,3,5,6-Tetrafluorophenol was used in preparation of radioiodinated phenylalanine derivative which is useful in peptide synthesis, technetium-99m labeled antibodies.

Solubility
Partly miscible with water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-39-1
Molecular Formula C6H2F4O
Molecular Weight (g/mol) 166.075
MDL Number MFCD00002157
InChI Key PBYIIRLNRCVTMQ-UHFFFAOYSA-N
Synonym phenol, 2,3,5,6-tetrafluoro, unii-tna118i5tg, 2,3,5,6-tetrafluoro phenol, tna118i5tg, 2,3,5,6-tetrafluor-phenol, ccris 5843, zlchem 411, pubchem2135, acmc-1bc5w, 2,3,5,6tetrafluorophenol
PubChem CID 69858
IUPAC Name 2,3,5,6-tetrafluorophenol
SMILES C1=C(C(=C(C(=C1F)F)O)F)F

Specifications

Melting Point 28°C to 30°C
Boiling Point 141°C to 143°C
Flash Point 79°C (174°F)
Odor Phenol-like
Quantity 5 g
UN Number UN3261
Beilstein 1911548
Solubility Information Partly miscible with water.
Formula Weight 166.07
Percent Purity 97%
Chemical Name or Material 2,3,5,6-Tetrafluorophenol
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 212-209-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.