Learn More
eMolecules​ 2,3,5,6-Tetramethylphenylboronic acid | 197223-36-2 | 5G | Purity: 96%
Supplier:  eMolecules​ 205382385
Encompass_Preferred
Combi-Blocks | 2,3,5,6-Tetramethylphenylboronic acid | 5G | 197223-36-2 | MFCD03093882
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-158-2901