Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole 98.0+%, TCI America™
SDP

Catalog No. H0718500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0718500G 500 g
H071825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0718500G Supplier TCI America Supplier No. H0718500G

Chemical Identifiers

CAS 25973-55-1
Molecular Formula C22H29N3O
Molecular Weight (g/mol) 351.49
MDL Number MFCD00134707
InChI Key ZMWRRFHBXARRRT-UHFFFAOYSA-N
PubChem CID 33263
IUPAC Name 2-(2H-1,2,3-benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC(N2N=C3C=CC=CC3=N2)=C(O)C(=C1)C(C)(C)CC

Specifications

Melting Point 82°C
Color Yellow
Quantity 500 g
Formula Weight 351.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole
EINECSNumber (5)-3604
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.