Learn More
Medchemexpress LLC 2-(3,5-difluoro-4-hydroxyanilino)-5,7-dimethyl-8-(3-methylbutyl)-7H-pteridin-6-one | 501437-28-1 | MFCD11223662 | 99.4% | 391.42 g·mol⁻1 | C19H23F2N5O2 | 100 MG
SDP
Catalog No. 5000243812
Encompass_Preferred
BI-D1870 is an ATP-competitive, cell-permeable, brain-penetrant small-molecule inhibitor of p90 ribosomal S6 kinases (RSK1-4) with low-nanomolar potency. It is intended for research use in biochemical and cellular studies targeting RSK-mediated signaling.
  • Inhibits RSK1-4 with reported IC50s of 31 nM, 24 nM, 18 nM, and 15 nM, respectively.
  • ATP-competitive mechanism of action.
  • Cell permeable and reported to be brain penetrant.
  • High chemical purity suitable for research (reported ≥99%).
  • Supplied as a solid and available as DMSO solutions for assays.
  • Molecular formula C19H23F2N5O2; molecular weight 391.42 g·mol⁻1.
Catalog No. 50-002-43812 Supplier Medchemexpress LLC Supplier No. HY10510100MG
BI-D1870 is an ATP-competitive, cell-permeable, brain-penetrant small-molecule inhibitor of p90 ribosomal S6 kinases (RSK1-4) with low-nanomolar potency. It is intended for research use in biochemical and cellular studies targeting RSK-mediated signaling.
  • Inhibits RSK1-4 with reported IC50s of 31 nM, 24 nM, 18 nM, and 15 nM, respectively.
  • ATP-competitive mechanism of action.
  • Cell permeable and reported to be brain penetrant.
  • High chemical purity suitable for research (reported ≥99%).
  • Supplied as a solid and available as DMSO solutions for assays.
  • Molecular formula C19H23F2N5O2; molecular weight 391.42 g·mol⁻1.