Learn More
Medchemexpress LLC 2-(3,5-difluoro-4-hydroxyanilino)-5,7-dimethyl-8-(3-methylbutyl)-7H-pteridin-6-one | 501437-28-1 | MFCD11223662 | 99.4% | 391.42 g·mol⁻¹ | C19H23F2N5O2 | 25 MG
SDP
Catalog No. 5000243954
Encompass_Preferred
BI-D1870 is an ATP-competitive, cell-permeable, brain-penetrant inhibitor of p90 ribosomal S6 kinases (RSK1-4) with low-nanomolar potency. It is used as a research tool in biochemical kinase assays and cell-based studies to probe RSK-dependent signaling pathways.
  • Low-nanomolar potency against RSK1-4 (reported IC50s ~31-15 nM).
  • ATP-competitive mechanism of inhibition, suitable for biochemical kinase assays.
  • Cell permeable and reported to penetrate the brain for in vivo applications.
  • High purity, typically around 99.4% for research-grade material.
  • Available in small pack sizes for laboratory use, e.g., 25 MG.
  • Well-characterized identifiers: CAS 501437-28-1, MFCD11223662.
Catalog No. 50-002-43954 Supplier Medchemexpress LLC Supplier No. HY1051025MG
BI-D1870 is an ATP-competitive, cell-permeable, brain-penetrant inhibitor of p90 ribosomal S6 kinases (RSK1-4) with low-nanomolar potency. It is used as a research tool in biochemical kinase assays and cell-based studies to probe RSK-dependent signaling pathways.
  • Low-nanomolar potency against RSK1-4 (reported IC50s ~31-15 nM).
  • ATP-competitive mechanism of inhibition, suitable for biochemical kinase assays.
  • Cell permeable and reported to penetrate the brain for in vivo applications.
  • High purity, typically around 99.4% for research-grade material.
  • Available in small pack sizes for laboratory use, e.g., 25 MG.
  • Well-characterized identifiers: CAS 501437-28-1, MFCD11223662.