Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3,5-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™
SDP

Catalog No. d35335g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35335G Supplier TCI America Supplier No. D35335G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 325142-93-6
Molecular Formula C14H21BO2
Molecular Weight (g/mol) 232.13
InChI Key XWCNEQGQJTWRHX-UHFFFAOYSA-N
Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-m-xylene, 3,5-Dimethylphenylboronic Acid Pinacol Ester
PubChem CID 15906198
IUPAC Name 2-(3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)C)C

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 232.13
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2-(3,5-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.