Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine Riboside 97.0+%, TCI America™
SDP

Catalog No. T26901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26901G Supplier TCI America Supplier No. T26901G

Chemical Identifiers

CAS 16321-99-6
Molecular Formula C16H18ClN5O7
Molecular Weight (g/mol) 427.80
MDL Number MFCD03425473
InChI Key HLVGVJFVAKPFOZ-UHFFFAOYNA-N
Synonym 2-Amino-6-chloro-9-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)purine
PubChem CID 10788628
IUPAC Name [3,4-bis(acetyloxy)-5-(2-amino-6-chloro-9H-purin-9-yl)oxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(C(OC(C)=O)C1OC(C)=O)N1C=NC2=C(Cl)N=C(N)N=C12

Specifications

Melting Point 148°C
Color Yellow
Quantity 1 g
Formula Weight 427.80
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′,5′-Tri-O-acetyl-2-amino-6-chloropurine Riboside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.