Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3',5'-Tri-O-acetyl-2-thiouridine | 28542-31-6 | 99.3% | 386.38 | C15H18N2O8S | 10 MG
SDP

Catalog No. 5000285948
Encompass_Preferred

2',3',5'-Tri-O-acetyl-2-thiouridine is an acetylated nucleoside analog supplied as a purified research reagent for biochemical and pharmacological studies. The material is provided as a solid, with documented analytical characterization and safety data for laboratory handling.

  • Acetylated nucleoside analog suitable for biochemical and pharmacological research.
  • High purity: 99.3%.
  • Chemical formula C15H18N2O8S; molecular weight 386.38.
  • CAS number 28542-31-6 for unambiguous identification.
  • Available in small-quantity vials (10 mg) for reagent use.
  • Includes certificate of analysis and safety data sheet for quality and handling.

Catalog No. 50-002-85948 Supplier Medchemexpress LLC Supplier No. HY15417010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',3',5'-Tri-O-acetyl-2-thiouridine is an acetylated nucleoside analog supplied as a purified research reagent for biochemical and pharmacological studies. The material is provided as a solid, with documented analytical characterization and safety data for laboratory handling.

  • Acetylated nucleoside analog suitable for biochemical and pharmacological research.
  • High purity: 99.3%.
  • Chemical formula C15H18N2O8S; molecular weight 386.38.
  • CAS number 28542-31-6 for unambiguous identification.
  • Available in small-quantity vials (10 mg) for reagent use.
  • Includes certificate of analysis and safety data sheet for quality and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.