Learn More
Medchemexpress LLC 2',3',5'-Tri-O-acetyl adenosine | 7387-57-7 | 97.0% | 393.35 | 10 G
SDP
Catalog No. 5000453170
Encompass_Preferred
2',3',5'-Tri-O-acetyl adenosine is an adenosine analog. Adenosine analogs primarily function as smooth muscle vasodilators and have also been shown to inhibit cancer progression.
  • Adenosine analog.
  • Acts as a smooth muscle vasodilator.
  • Inhibits cancer progression.
Catalog No. 50-004-53170 Supplier Medchemexpress LLC Supplier No. HYW00901610G
2',3',5'-Tri-O-acetyl adenosine is an adenosine analog. Adenosine analogs primarily function as smooth muscle vasodilators and have also been shown to inhibit cancer progression.
  • Adenosine analog.
  • Acts as a smooth muscle vasodilator.
  • Inhibits cancer progression.