Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3',5'-Tri-O-acetyl adenosine | 7387-57-7 | 99.6% | 393.35 | 100 G
SDP

Catalog No. 5000451221
Encompass_Preferred

2',3',5'-Tri-O-acetyl adenosine is an adenosine analog that primarily acts as a smooth muscle vasodilator and has been shown to inhibit cancer progression. This product is suitable for laboratory use and storage, with specific conditions to maintain its stability.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, protect from light
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-51221 Supplier Medchemexpress LLC Supplier No. HYW009016100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',3',5'-Tri-O-acetyl adenosine is an adenosine analog that primarily acts as a smooth muscle vasodilator and has been shown to inhibit cancer progression. This product is suitable for laboratory use and storage, with specific conditions to maintain its stability.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, protect from light
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.