Learn More
Medchemexpress LLC 2',3',5'-Tri-O-acetyl adenosine | 7387-57-7 | 99.6% | 393.35 | 5 G
SDP
Catalog No. 5000453140
Encompass_Preferred
2',3',5'-Tri-O-acetyl adenosine is an adenosine analog that primarily acts as a smooth muscle vasodilator and has been shown to inhibit cancer progression. It is intended for research use only.
  • Adenosine analog
  • Smooth muscle vasodilator
  • Inhibits cancer progression
  • Solid appearance
  • White to off-white color
  • Store at -20°C, protect from light
Catalog No. 50-004-53140 Supplier Medchemexpress LLC Supplier No. HYW0090165G
2',3',5'-Tri-O-acetyl adenosine is an adenosine analog that primarily acts as a smooth muscle vasodilator and has been shown to inhibit cancer progression. It is intended for research use only.
  • Adenosine analog
  • Smooth muscle vasodilator
  • Inhibits cancer progression
  • Solid appearance
  • White to off-white color
  • Store at -20°C, protect from light