Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Tri-O-Benzoyl-1-O-Acetyl-β-D-Ribofuranose, MP Biomedicals

Catalog No. ICN15395525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15395525 25 g
ICN15395580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15395525 Supplier MP Biomedicals, Inc Supplier No. 0215395525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6974-32-9
Molecular Formula C28H24O9
Molecular Weight (g/mol) 504.491
InChI Key GCZABPLTDYVJMP-CBUXHAPBSA-N
Synonym 1-o-acetyl-2,3,5-tri-o-benzoyl-beta-d-ribofuranose, beta-d-ribofuranose 1-acetate 2,3,5-tribenzoate, 2r,3r,4r,5s-5-acetyloxy-3,4-bis benzoyloxy oxolan-2-yl methyl benzoate, 1-acetyl-2,3,5-tribenzoyl-beta-d-ribofuranose, 1-acetyl-2,3,5-tribenzoy-b-d-ribofuranose, 1-o-acetyl-2,3,5,tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5s-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl methyl benzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxymethyl tetrahydrofuran-3,4-diyl dibenzoate, .beta.-d-ribofuranose, 1-acetate 2,3,5-tribenzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxy methyl tetrahydrofuran-3,4-diyl dibenzoate
PubChem CID 81455
IUPAC Name [(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
SMILES CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

Color White
Quantity 25 g
Formula Weight 504.5
Physical Form Free-flowing Powder
Chemical Name or Material 2, 3, 5-Tri-O-Benzoyl-1-O-Acetyl-β-D-Ribofuranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.