Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-arabinofuranose, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6616506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6616506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6616506 Supplier Thermo Scientific Chemicals Supplier No. J6616506
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 52522-49-3 | Molecular Formula: C33H31NO8 | Molecular Weight: 569.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52522-49-3
Molecular Formula C33H31NO8
Molecular Weight (g/mol) 569.61
MDL Number MFCD00038477
InChI Key VBDBDZHLJKDSSB-UFRUDBOASA-N
Synonym 2,3,5-tri-o-benzyl-1-o-4-nitrobenzoyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-1-o-p-nitrobenzoyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-d-arabinofuranose 1-4-nitrobenzoate, d-arabinofuranose,2,3,5-tris-o-phenylmethyl-, 4-nitrobenzoate 9ci, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-yl 4-nitrobenzoate, 3s,4s,5r-3,4-bis phenylmethoxy-5-phenylmethoxy methyl oxolan-2-yl 4-nitro benzoate, 1-o-4-nitrobenzoyl-2,3,5-tri-o-benzyl-d-arabinofuranose, 4-nitrobenzoate 2,3,5-tris-o-benzyl-d-arabinofuranose, 3s,4r,5r-3,4-dibenzyloxy-5-benzyloxymethyl tetrahydrofuran-2-yl 4-nitrobenzoate
PubChem CID 97076
IUPAC Name [(3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-yl] 4-nitrobenzoate
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)OC(=O)C3=CC=C(C=C3)[N+](=O)[O-])OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 69°C to 72°C
Color White
Quantity 5 g
Beilstein 1303859
Formula Weight 569.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-arabinofuranose
EINECSNumber 257-992-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.