Learn More
Medchemexpress LLC 2,3,5-tri-O-benzyl-D-ribose | 54623-25-5 | MFCD03425640 | >95.0% | 420.5 g/mol | C26H28O5 | 5 G
SDP
Catalog No. 5000300366
Encompass_Preferred
2,3,5-Tri-O-benzyl-D-ribose is a benzyl-protected derivative of D-ribose used as a synthetic intermediate in carbohydrate and nucleoside chemistry. It protects hydroxyl groups to enable selective transformations and the assembly of complex molecules while being compatible with common organic reaction conditions.
  • Benzyl-protected sugar suitable for multi-step synthesis.
  • Facilitates selective functionalization and coupling reactions.
  • Soluble in common organic solvents for easy handling.
  • Stable under recommended storage conditions to preserve integrity.
  • Characterized identity including CAS and molecular data for traceability.
Catalog No. 50-003-00366 Supplier Medchemexpress LLC Supplier No. HYW0372005G
2,3,5-Tri-O-benzyl-D-ribose is a benzyl-protected derivative of D-ribose used as a synthetic intermediate in carbohydrate and nucleoside chemistry. It protects hydroxyl groups to enable selective transformations and the assembly of complex molecules while being compatible with common organic reaction conditions.
  • Benzyl-protected sugar suitable for multi-step synthesis.
  • Facilitates selective functionalization and coupling reactions.
  • Soluble in common organic solvents for easy handling.
  • Stable under recommended storage conditions to preserve integrity.
  • Characterized identity including CAS and molecular data for traceability.