Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Tribromo-4-methylthiophene 97.0+%, TCI America™
SDP

Catalog No. T168210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T168210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T168210G Supplier TCI America Supplier No. T168210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67869-13-0
Molecular Formula C5H3Br3S
Molecular Weight (g/mol) 334.851
MDL Number MFCD00130086
InChI Key DTLNXOZXOZCNMX-UHFFFAOYSA-N
PubChem CID 3017801
IUPAC Name 2,3,5-tribromo-4-methylthiophene
SMILES CC1=C(SC(=C1Br)Br)Br

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 334.85
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,5-Tribromo-4-methylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.