Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Tribromopyridine 98.0+%, TCI America™
SDP

Catalog No. T33395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33395G 5 g
T333925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33395G Supplier TCI America Supplier No. T33395G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75806-85-8
Molecular Formula C5H2Br3N
Molecular Weight (g/mol) 315.79
MDL Number MFCD00233999
InChI Key BBHYMJRPJFVNPI-UHFFFAOYSA-N
PubChem CID 628798
IUPAC Name 2,3,5-tribromopyridine
SMILES C1=C(C=NC(=C1Br)Br)Br

Specifications

Melting Point 46°C
Boiling Point 160°C
Quantity 5 g
Formula Weight 315.79
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5-Tribromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.