Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Tribromothiophene, 99+%, Thermo Scientific™

Catalog No. AC259840010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC259840010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259840010 Supplier Thermo Scientific Chemicals Supplier No. 259840010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3141-24-0
Molecular Formula C4HBr3S
Molecular Weight (g/mol) 320.82
MDL Number MFCD00014521
InChI Key SKDNDSLDRLEELJ-UHFFFAOYSA-N
Synonym thiophene, 2,3,5-tribromo, pubchem5608, 2,5-tribromothiophene, acmc-209hmg, thiophene,3,5-tribromo, 2,3,5-tribromo thiophene, ksc225k9l, 2,3,5-tribromothiophene
PubChem CID 76592
IUPAC Name 2,3,5-tribromothiophene
SMILES BrC1=CC(Br)=C(Br)S1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 27°C to 29°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 1 g
Formula Weight 320.82
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3, 5-Tribromothiophene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.