Learn More
Medchemexpress LLC 2 3 5-Trichloro-4-py 50mg
SDP
Catalog No. 5000204905
Encompass_Preferred
2 3 5-Trichloro-4-pyridinol can be used as a herbicide for direct-seeded and flooded rice[1]
Catalog No. 50-002-04905 Supplier Medchemexpress LLC Supplier No. HYW10523350MG
2 3 5-Trichloro-4-pyridinol can be used as a herbicide for direct-seeded and flooded rice[1]