Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trifluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T23855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23855G 5 g
T23851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23855G Supplier TCI America Supplier No. T23855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 654-87-5
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.09
MDL Number MFCD00061200
InChI Key CPZROMDDCPPFOO-UHFFFAOYSA-N
Synonym 2,3,5-trilfluorobenzoic acid, benzoic acid, 2,3,5-trifluoro, pubchem1401, acmc-209nr1, 2,3,5-trifluorobenzoicacid, ksc622o6j, 2,3,5-trifluoro-benzoic acid, rarechem al bo 0710, benzoic acid,2,3,5-trifluoro, cpzromddcppfoo-uhfffaoysa
PubChem CID 2776991
IUPAC Name 2,3,5-trifluorobenzoic acid
SMILES OC(=O)C1=CC(F)=CC(F)=C1F

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 5 g
Formula Weight 176.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5-Trifluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.