Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trifluorobenzonitrile 97.0+%, TCI America™
SDP

Catalog No. T23695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23695G Supplier TCI America Supplier No. T23695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 241154-09-6
Molecular Formula C7H2F3N
Molecular Weight (g/mol) 157.095
MDL Number MFCD00083539
InChI Key MMPKVFQMDMBXSG-UHFFFAOYSA-N
Synonym benzonitrile, 2,3,5-trifluoro, 2,3,5-trifluorobenzenecarbonitrile, pubchem4823, intermediates-zcf02648, ksc552s4h, 2,3,5-trifluoro-benzonitrile, benzonitrile,2,3,5-trifluoro, 2,3,5-trifluoro benzonitrile
PubChem CID 2776998
IUPAC Name 2,3,5-trifluorobenzonitrile
SMILES C1=C(C=C(C(=C1F)F)C#N)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3276
Formula Weight 157.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5-Trifluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.