Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3,5-Trifluorobenzylamine

Catalog No. CDS002986 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002986 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002986 Supplier Sigma Aldrich Supplier No. CDS002986250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 4-methyl-4-pentenoate is an olefin ester
  • It undergoes iron carbonyl-promoted isomerization to α,α-unsaturated esters
  • Reaction of ethyl 4-methyl-4-pentenoate with ch3oboch3+ was examined in a small fourier-transform ion cyclotron resonance mass spectrometer equipped with a permanent magnet

Specifications

CAS 244022-72-8
Molecular Formula C7H6F3N
Linear Formula C7H6F3N
MDL Number MFCD00236314
Quantity 5 g
Molecular Weight (g/mol) 161.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.