Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trimethylanisole 97.0+%, TCI America™
SDP

Catalog No. T24565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24565G 5 g
T245625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24565G Supplier TCI America Supplier No. T24565G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20469-61-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00142783
InChI Key AWONIZVBKXHWJP-UHFFFAOYSA-N
Synonym 2,3,5-trimethylanisole, anisole, 2,3,5-trimethyl, benzene, 1-methoxy-2,3,5-trimethyl, 1-methoxy-2,3,5-trimethyl-benzene, pubchem2688, acmc-209fam, 2,3,5-trimethylanizole, 3-methoxy-1,2,5-trimethylbenzene
PubChem CID 88555
IUPAC Name 1-methoxy-2,3,5-trimethylbenzene
SMILES COC1=CC(C)=CC(C)=C1C

Specifications

Color Yellow
Boiling Point 216°C
Quantity 5 g
Formula Weight 150.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5-Trimethylanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.