Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trimethylpyrazine 98.0+%, TCI America™
SDP

Catalog No. T094225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T094225G 25 g
T09425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T094225G Supplier TCI America Supplier No. T094225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14667-55-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006145
InChI Key IAEGWXHKWJGQAZ-UHFFFAOYSA-N
Synonym trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
PubChem CID 26808
IUPAC Name 2,3,5-trimethylpyrazine
SMILES CC1=CN=C(C(=N1)C)C

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3,5-Trimethylpyrazine
RTECSNumber UQ3907000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.