Learn More
2,3,5-Triphenyl-2H-tetrazolium Chloride Monohydrate,MP Biomedicals
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| Color | Tan |
| Quantity | 50 g |
| Formula Weight | 334.81 |
| Physical Form | Powder |
| Chemical Name or Material | 2, 3, 5-Triphenyl-2H-tetrazolium Chloride Monohydrate |
Safety and Handling
| EINECSNumber | 206-071-6 |
| storageNote1 | Protect from light. |
| Recommended Storage | Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.