Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Triphenyl-2H-tetrazolium Chloride Monohydrate,MP Biomedicals

Catalog No. ICN229893 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN229893 10 g
ICN10312650 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN229893 Supplier MP Biomedicals, Inc Supplier No. 0522989310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 298-96-4
Molecular Formula C19H15ClN4
Molecular Weight (g/mol) 334.81
MDL Number MFCD00011963
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
Synonym 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride
PubChem CID 9283
ChEBI CHEBI:78019
IUPAC Name triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Tan
Quantity 10 g
Formula Weight 334.81
Physical Form Powder
Chemical Name or Material 2, 3, 5-Triphenyl-2H-tetrazolium Chloride Monohydrate
EINECSNumber 206-071-6
storageNote1 Protect from light.
Recommended Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.