Learn More
Sigma Aldrich Fine Chemicals Biosciences 2,3,5-Triphenyltetrazolium chloride, 298-96-4, MFCD00011963, 5 G
Catalog No. NC0578152
Encompass
Empirical Formula (Hill Notation): C19H15ClN4, Molecular Weight: 334.80, ≥98.0% (HPLC), Synonym: TPTZ, TTZ, Tetrazolium Red. 2,3,5-Triphenyltetrazolium chloride has been used to stain heart tissue to measure the extent of acute lesion. It has also been used to stain brain tissue to check the size of infarct area.
Catalog No. NC0578152 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T88775G
Empirical Formula (Hill Notation): C19H15ClN4, Molecular Weight: 334.80, ≥98.0% (HPLC), Synonym: TPTZ, TTZ, Tetrazolium Red. 2,3,5-Triphenyltetrazolium chloride has been used to stain heart tissue to measure the extent of acute lesion. It has also been used to stain brain tissue to check the size of infarct area.