Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Triphenyltetrazolium Chloride, Sigma-Aldrich

Catalog No. 01258827 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
50 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-827 5 g
01-258-877 10 g
01-258-828 25 g
01-258-878 50 g
01-258-826 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-258-827 Supplier MilliporeSigma Supelco Supplier No. T88775G
Add to Cart
Add to Cart

2,3,5-Triphenyltetrazolium chloride has been used to stain heart tissue to measure the extent of acute lesion. It has also been used to stain brain tissue to check the size of infarct area.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-96-4

Specifications

Quantity 5 g
Molecular Formula C19H15ClN4
Form Powder
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol) 334.81, 334.8 g/mol
Concentration ≥98%
Product Type Differentiator Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.