Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6,7,10,11-Hexabromotriphenylene 98.0+%, TCI America™
SDP

Catalog No. H1201100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1201100MG 100 mg
H12011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1201100MG Supplier TCI America Supplier No. H1201100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82632-80-2
Molecular Formula C18H6Br6
Molecular Weight (g/mol) 701.67
InChI Key GLHQUXLCQLQNPZ-UHFFFAOYSA-N
PubChem CID 11083045
IUPAC Name 2,3,6,7,10,11-hexabromotriphenylene
SMILES C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1Br)Br)Br)Br)Br)Br

Specifications

Quantity 100 mg
Formula Weight 701.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6,7,10,11-Hexabromotriphenylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.