Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6-Trifluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAL2005303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2005303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2005303 Supplier Thermo Scientific Chemicals Supplier No. L2005303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 84940-46-5 | Molecular Formula: C6HF6N | Molecular Weight: 201.071 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84940-46-5
Molecular Formula C6HF6N
Molecular Weight (g/mol) 201.071
MDL Number MFCD03840203
InChI Key DODFHNWFNCGXOW-UHFFFAOYSA-N
Synonym 2,3,6-trifluoro-4-trifluoromethyl pyridine, pyridine,2,3,6-trifluoro-4-trifluoromethyl, 2,3,6-tris fluoranyl-4-trifluoromethyl pyridine
PubChem CID 2782842
IUPAC Name 2,3,6-trifluoro-4-(trifluoromethyl)pyridine
SMILES C1=C(C(=C(N=C1F)F)F)C(F)(F)F

Specifications

Density 1.579
Boiling Point 104°C to 106°C
Refractive Index 1.3774
Quantity 1 g
UN Number UN1993
Beilstein 6399655
Formula Weight 201.07
Percent Purity 97%
Chemical Name or Material 2,3,6-Trifluoro-4-(trifluoromethyl)pyridine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.