Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6-Trifluorotoluene, 98%, Thermo Scientific™

Catalog No. AAH6389703
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6389703 1 g
AAH63897MD 250 mg
AAH6389706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6389703 Supplier Thermo Scientific Chemicals Supplier No. H6389703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119916-25-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD12922564
InChI Key UZYYBZNZSSNYSA-UHFFFAOYSA-N
Synonym 2,3,6-trichlorotoluene, benzene, 1,2,4-trichloro-3-methyl, toluene, 2,3,6-trichloro, unii-19m2l15z6u, 2,3,6-trichorotoluene, 2,6-trichlorotoluene, toluene,3,6-trichloro, 2,3,6-trichloro-toluen, toluene,2,3,6-trichloro, 4-05-00-00819 beilstein handbook reference
PubChem CID 16378
IUPAC Name 1,2,4-trichloro-3-methylbenzene
SMILES CC1=C(C=CC(=C1Cl)Cl)Cl

Specifications

Color Colorless to Yellow
Quantity 1 g
Physical Form Liquid
Formula Weight 146.11
Percent Purity 98%
Chemical Name or Material 2,3,6-Trifluorotoluene
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.