Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione 98.0+%, TCI America™
SDP

Catalog No. T26351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26351G 1 g
T26355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26351G Supplier TCI America Supplier No. T26351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19576-08-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00135278
InChI Key FNYAZSZTENLTRT-UHFFFAOYSA-N
Synonym Hajos-Parrish Ketone
PubChem CID 596311
IUPAC Name 7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione
SMILES CC12CCC(=O)C=C1CCC2=O

Specifications

Melting Point 71°C
Quantity 1 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.