Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine copper(II)

Catalog No. 257540100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-754-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-754-0100MG Supplier Sigma Aldrich Supplier No. 257540100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine metalates with copper (II) to form 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine copper(II). CuOEP was used to prepare a mixed adlayer with cobalt phthalocyanine, by immersing an Au(111) substrate.

Specifications

CAS 14409-63-3
Molecular Formula C36H44CuN4
Linear Formula C36H44CuN4
Quantity 100 mg
MDL Number MFCD00012150
Molecular Weight (g/mol) 596.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.