Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine palladium(II)

Catalog No. 392014100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-201-4100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-201-4100MG Supplier Sigma Aldrich Supplier No. 392014100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine palladium (II) can be formed by metalating 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine with palladium (II).1

Specifications

CAS 24804-00-0
Molecular Formula C36H44N4Pd
Linear Formula C36H44N4Pd
MDL Number MFCD00192364
Quantity 100 mg
Molecular Weight (g/mol) 639.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.