Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,7,8,12,13,17,18-Octaethylporphyrin 95.0+%, TCI America™
SDP

Catalog No. O0234100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0234100MG 100 mg
O02341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0234100MG Supplier TCI America Supplier No. O0234100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2683-82-1
Molecular Formula C36H46N4
Molecular Weight (g/mol) 534.792
MDL Number MFCD00009627
InChI Key XFIIGRBIXXECHR-UHFFFAOYSA-N
Synonym octaethylporphine, octaethylporphyrin, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethylporphyrin, octaethylpophine, 21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethyl, octaethylporphyrine, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphin, acmc-209gte, hciifbhdbocsaf-muzkialcsa-n
PubChem CID 102311
IUPAC Name 2,3,7,8,12,13,17,18-octaethyl-21,22-dihydroporphyrin
SMILES CCC1=C(C2=CC3=NC(=CC4=NC(=CC5=C(C(=C(N5)C=C1N2)CC)CC)C(=C4CC)CC)C(=C3CC)CC)CC

Specifications

Quantity 100 mg
Formula Weight 534.79
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,7,8,12,13,17,18-Octaethylporphyrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.