Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,7,8-Tetrachlorodibenzofuran Solution, 50μg/mL in Toluene, Ultra Scientific

Catalog No. USRPE037S
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPE-037S 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-RPE-037S Supplier Agilent Technologies Supplier No. RPE037S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2050-67-1 , 108-88-3
Molecular Formula C12H8Cl2
Molecular Weight (g/mol) 223.096
InChI Key KTXUOWUHFLBZPW-UHFFFAOYSA-N
PubChem CID 16307
IUPAC Name 1-chloro-3-(3-chlorophenyl)benzene
SMILES C1=CC(=CC(=C1)Cl)C2=CC(=CC=C2)Cl

Specifications

Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 50μg/mL
Chemical Name or Material 2,3,7,8-Tetrachlorodibenzofuran Solution, 50μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.