Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(3-Acetylphenoxy)-N-phenylacetamide

Catalog No. OTV000479 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000479 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000479 Supplier Sigma Aldrich Supplier No. OTV000479250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,5-dimethoxytoluene (dmt) is a methoxylated phenolic derivative
  • It is reported to be one of the main constituent of the floral volatiles in different rose varieties
  • It has been biosynthesized from orcinol by two successive methylation catalyzed by o-methyltransferases (omts)
  • The features of its aerobic oxidation with metal/bromide catalysts have been investigated.3,5-dimethoxytoluene (dmt) may be used in the synthesis of 3,5-dimethoxybenzoic acid by oxidation and 2-methoxy-6-methyl-1,4-benzoquinone by catalytic oxidation with hydrogen peroxide (h2o2)/methyltrioxorhenium (ch3reo3) in dimethyl carbonate (dmc)

Specifications

Molecular Formula C16H15NO3
Linear Formula C16H15NO3
MDL Number MFCD04790713
Quantity 1 g
Molecular Weight (g/mol) 269.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.