Learn More
Medchemexpress LLC 2-(3-amino-6-chloroquinolin-2-yl)propan-2-ol | 916056-79-6 | MFCD28502442 | 98.5% | 236.70 | C12H13ClN2O | 25 MG
SDP
Catalog No. 5000239716
Encompass_Preferred
Reproxalap is a reactive aldehyde-sequestering small molecule used in research to study ocular inflammation, including dry eye disease, allergic conjunctivitis, and non-infectious anterior uveitis. It reduces PKCα activity, blocks caspase 3/7 activation, and protects cells from aldehyde-induced cytotoxicity.
  • Reactive aldehyde sequestering mechanism for cellular protection.
  • Reported purity of 98.5% suitable for research use.
  • Solid powder form with recommended refrigerated or frozen storage for stability.
  • Molecular formula C12H13ClN2O; molecular weight 236.70 for identification.
  • Intended for research use only, not for human therapeutic use.
Catalog No. 50-002-39716 Supplier Medchemexpress LLC Supplier No. HY10715025MG
Reproxalap is a reactive aldehyde-sequestering small molecule used in research to study ocular inflammation, including dry eye disease, allergic conjunctivitis, and non-infectious anterior uveitis. It reduces PKCα activity, blocks caspase 3/7 activation, and protects cells from aldehyde-induced cytotoxicity.
  • Reactive aldehyde sequestering mechanism for cellular protection.
  • Reported purity of 98.5% suitable for research use.
  • Solid powder form with recommended refrigerated or frozen storage for stability.
  • Molecular formula C12H13ClN2O; molecular weight 236.70 for identification.
  • Intended for research use only, not for human therapeutic use.