Learn More
Medchemexpress LLC 2-(3-amino-6-chloroquinolin-2-yl)propan-2-ol | 916056-79-6 | MFCD28502442 | 98.5% | 236.70 g·mol⁻¹ | C12H13ClN2O | 100 MG
SDP
Catalog No. 5000239778
Encompass_Preferred
Reproxalap (ADX-102) is a reactive aldehyde species (RASP) sequestering small molecule used in preclinical research into ocular inflammation and related conditions. It reduces PKCα activity, inhibits caspase 3/7 activation, and protects cells from aldehyde-induced cytotoxicity.
  • Reactive aldehyde sequestering mechanism
  • Reduces PKCα activity and blocks caspase 3/7 activation
  • High purity: 98.49%
  • Solid powder form suitable for formulation and dosing
  • Stable under recommended storage conditions
  • Available in 100 mg pack size
Catalog No. 50-002-39778 Supplier Medchemexpress LLC Supplier No. HY107150100MG
Reproxalap (ADX-102) is a reactive aldehyde species (RASP) sequestering small molecule used in preclinical research into ocular inflammation and related conditions. It reduces PKCα activity, inhibits caspase 3/7 activation, and protects cells from aldehyde-induced cytotoxicity.
  • Reactive aldehyde sequestering mechanism
  • Reduces PKCα activity and blocks caspase 3/7 activation
  • High purity: 98.49%
  • Solid powder form suitable for formulation and dosing
  • Stable under recommended storage conditions
  • Available in 100 mg pack size