Learn More
eMolecules​ 88192-21-6 | 2-(3-azidopropyl)-isoindole-1,3-dione | Synthonix - Stock | MFCD13480971 | 230.227 | C11H10N4O2 | 97.000 | [N-]=[N+]=NCCCN1C(=O)c2ccccc2C1=O | 10g | 492245362
Catalog No. 502697273 Shop All eMolecules Inc. Products
Encompass_Preferred
2-(3-azidopropyl)-isoindole-1,3-dione | Synthonix - Stock | 88192-21-6 | MFCD13480971 | 230.227 | C11H10N4O2 | 97.000 | [N-]=[N+]=NCCCN1C(=O)c2ccccc2C1=O | 10g | 492245362
Catalog No. 50-269-7273 Supplier eMolecules​ Supplier No. 492245362
2-(3-azidopropyl)-isoindole-1,3-dione | Synthonix - Stock | 88192-21-6 | MFCD13480971 | 230.227 | C11H10N4O2 | 97.000 | [N-]=[N+]=NCCCN1C(=O)c2ccccc2C1=O | 10g | 492245362